siRNA Details
| Gene Symbol: Ccdc88a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176841.4 | NP_789811.2 | 20, 21 | 3784 | ||
| GenBank mRNA | AB087827.1 | - | 20, 21 | 3608 | ||
| AK020777.1 | - | 2, 3 | 389 | |||
| AK129310.1 | - | 7, 8 | 1772 | |||
| BC037020.1 | - | 2, 3 | 353 | |||
| BC079895.1 | - | 11, 12 | 2630 | |||
| CN458498.1 | - | 4, 5 | 679 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008770470, FQ230491.

Ccdc88a
Genomic Map