siRNA Details
| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176844.4 | NP_789814.2 | 3, 4 | 438 | ||
| GenBank mRNA | AF204689.1 | - | 2, 3 | 203 | ||
| AY574264.1 | - | 3, 4 | 221 | |||
| BC106839.2 | - | 3, 4 | 221 | 40045716 | ||
| BC106853.2 | - | 3, 4 | 221 | 40046382 | ||
| DQ318788.1 | - | 3, 4 | 391 | |||

Chrna5
Genomic Map