siRNA Details
| Gene Symbol: Vps37d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199677.1 | NP_001186606.1 | 4 | 1139 | ||
| NM_177574.4 | NP_808242.1 | 2 | 884 | |||
| GenBank mRNA | AK090181.1 | - | 2 | 886 | ||
| AK158287.1 | - | 4 | 1068 | |||
| AY081953.2 | - | 4 | 1139 | |||
| AY352519.1 | - | 2 | 884 | |||
| BC044789.1 | - | 4 | 1042 | 3484221 | ||

Vps37d
Genomic Map