siRNA Details
| Gene Symbol: Dcdc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195617.1 | NP_001182546.1 | 2, 3 | 609 | ||
| NM_177577.3 | NP_808245.2 | 2, 3 | 609 | |||
| GenBank mRNA | AK220370.1 | - | 2, 3 | 382 | ||
| BC045136.1 | - | 2, 3 | 609 | 6306251 | ||

Dcdc2a
Genomic Map