siRNA Details
| Gene Symbol: Btla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037719.2 | NP_001032808.2 | 2 | 361 | ||
| NM_177584.3 | NP_808252.1 | 2 | 361 | |||
| GenBank mRNA | AK041334.1 | - | 2 | 363 | ||
| AY293285.1 | - | 2 | 345 | |||
| BC108963.1 | - | 2 | 363 | 40046744 | ||
| BC108964.1 | - | 2 | 363 | 40046745 | ||

Btla
Genomic Map