siRNA Details
Gene Symbol: Zfp369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178364.5 | NP_848141.3 | 8 | 1874 | ||
GenBank mRNA | AJ319726.1 | - | 8 | 1891 | ||
AK084309.1 | - | 8 | 1877 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178364.5 | NP_848141.3 | 8 | 1874 | ||
GenBank mRNA | AJ319726.1 | - | 8 | 1891 | ||
AK084309.1 | - | 8 | 1877 |