siRNA Details
| Gene Symbol: CKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001824.4 | NP_001815.2 | 2 | 318 | ||
| XM_017026240.1 | XP_016881729.1 | 1 | 203 | |||
| GenBank mRNA | AK129878.1 | - | 2 | 216 | ||
| AK300730.1 | - | 2 | 216 | |||
| AK313281.1 | - | 2 | 216 | |||
| BC007462.1 | - | 2 | 216 | 3934232 | ||
| BT006793.1 | - | 1 | 143 | |||
| DA896166.1 | - | 2 | 316 | |||
| DC385783.1 | - | 2 | 319 | |||

CKM
Genomic Map