siRNA Details
| Gene Symbol: Rbms3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172121.1 | NP_001165592.1 | 6 | 698 | ||
| NM_001172122.1 | NP_001165593.1 | 5 | 1170 | |||
| NM_001172123.1 | NP_001165594.1 | 5 | 1170 | |||
| NM_001172124.1 | NP_001165595.1 | 5 | 1170 | |||
| NM_001172126.1 | NP_001165597.1 | 5 | 1170 | |||
| NM_178660.4 | NP_848775.2 | 5 | 1170 | |||
| GenBank mRNA | AK030478.1 | - | 5 | 830 | ||
| AK030609.1 | - | 5 | 804 | |||
| AK032938.1 | - | 5 | 1167 | |||
| AK033005.1 | - | 5 | 813 | |||
| AK041098.1 | - | 5 | 1172 | |||
| AK046197.1 | - | 6 | 699 | |||
| AK051776.1 | - | 6 | 1217 | |||
| AK052196.1 | - | 5 | 1151 | |||
| BC033604.1 | - | 1 | 25 | |||
| BC108402.1 | - | 5 | 1144 | 6516420 | ||
| BC117841.1 | - | 5 | 610 | 40093225 | ||

Rbms3
Genomic Map