siRNA Details
| Gene Symbol: Bend7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190400.1 | NP_001177329.1 | 9 | 1974 | ||
| NM_178663.4 | NP_848778.1 | 9 | 1977 | |||
| GenBank mRNA | AK030428.1 | - | 9 | 1664 | ||
| AK053896.1 | - | 9 | 1976 | |||
| BC056476.1 | - | 6 | 1021 | |||
| BC096430.1 | - | 9 | 1646 | 5041388 | ||

Bend7
Genomic Map