siRNA Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289572.1 | NP_001276501.1 | 5 | 752 | ||
| NM_001289574.1 | NP_001276503.1 | 6 | 914 | |||
| NM_178914.4 | NP_849245.2 | 6 | 848 | |||
| GenBank mRNA | AK036359.1 | - | 6 | 832 | ||
| AK077500.1 | - | 6 | 759 | |||
| AK161308.1 | - | 5 | 698 | |||
| BC046960.1 | - | 6 | 806 | 6391450 | ||
| DQ196101.1 | - | 5 | 578 | |||

Spata7
Genomic Map