siRNA Details
| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178929.4 | NP_849260.2 | 5 | 1256 | ||
| GenBank mRNA | AK030678.1 | - | 5 | 1255 | ||
| AK155095.1 | - | 6 | 1501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kazald1| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178929.4 | NP_849260.2 | 5 | 1256 | ||
| GenBank mRNA | AK030678.1 | - | 5 | 1255 | ||
| AK155095.1 | - | 6 | 1501 | |||