siRNA Details
| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180600.3 | NP_850931.2 | 10 | 1184 | ||
| GenBank mRNA | AK076148.1 | - | 10 | 1047 | ||
| AK083216.1 | - | 10 | 1177 | |||
| AK150776.1 | - | 10 | 1009 | |||
| BC029111.1 | - | 10 | 1013 | 2631258 | ||

Ube2q2
Genomic Map