siRNA Details
| Gene Symbol: Vps37c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181403.2 | NP_852068.1 | 5 | 1561 | ||
| GenBank mRNA | AK030717.1 | - | 4 | 1432 | ||
| AK150606.1 | - | 5 | 1542 | |||
| AK158833.1 | - | 4 | 1300 | |||
| AK159309.1 | - | 5 | 1634 | |||
| AK165826.1 | - | 5 | 1629 | |||
| AK189300.1 | - | 1 | 248 | |||
| AK203174.1 | - | 1 | 129 | |||
| BC025865.1 | - | 5 | 1561 | 5029143 | ||

Vps37c
Genomic Map