siRNA Details
| Gene Symbol: Kank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181404.5 | NP_852069.4 | 5 | 2539 | ||
| GenBank mRNA | AK147945.1 | - | 2 | 2127 | ||
| BC058673.1 | - | 2 | 323 | |||
| BC079563.1 | - | 1 | 1866 | 30535512 | ||
| BC152400.1 | - | 1 | 841 | |||

Kank1
Genomic Map