siRNA Details
| Gene Symbol: Mtmr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181409.3 | NP_852074.2 | 15 | 1766 | ||
| GenBank mRNA | AK132375.1 | - | 15 | 1733 | ||
| BC042764.1 | - | 3 | 286 | |||
| BC051083.1 | - | 15 | 1699 | 6486043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr11| Gene Symbol: Mtmr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181409.3 | NP_852074.2 | 15 | 1766 | ||
| GenBank mRNA | AK132375.1 | - | 15 | 1733 | ||
| BC042764.1 | - | 3 | 286 | |||
| BC051083.1 | - | 15 | 1699 | 6486043 | ||