siRNA Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181728.2 | NP_859417.2 | 4 | 925 | ||
| GenBank mRNA | AJ311769.1 | - | 2 | 709 | ||
| AJ311770.1 | - | 2 | 709 | |||
| AJ311771.1 | - | 2 | 709 | |||
| AJ311772.1 | - | 2 | 709 | |||
| AJ311773.1 | - | 2 | 709 | |||
| AK016257.1 | - | 4 | 918 | |||
| BC141069.1 | - | 2 | 785 | 9055681 | ||
| BC145327.1 | - | 2 | 785 | 9053883 | ||
| BE625316.1 | - | 1 | 50 | |||

Art3
Genomic Map