siRNA Details
| Gene Symbol: Fgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181849.2 | NP_862897.1 | 3 | 369 | ||
| GenBank mRNA | AK011118.1 | - | 3 | 389 | ||
| AK011920.1 | - | 3 | 385 | |||
| AK146960.1 | - | 3 | 389 | |||
| AK168626.1 | - | 3 | 383 | |||
| BC031715.1 | - | 3 | 369 | 4195310 | ||

Fgb
Genomic Map