siRNA Details
| Gene Symbol: Magea9-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC116352.1 | - | 1 | 309 | ||
| BC116353.1 | - | 1 | 311 | 40099901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magea9-ps| Gene Symbol: Magea9-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC116352.1 | - | 1 | 309 | ||
| BC116353.1 | - | 1 | 311 | 40099901 | ||