siRNA Details
| Gene Symbol: Rnf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163263.1 | NP_001156735.1 | 20 | 3443 | ||
| NM_182999.2 | NP_892044.1 | 21 | 3290 | |||
| GenBank mRNA | AK220492.1 | - | 20 | 3359 | ||
| BC052482.1 | - | 21 | 3229 | 6308280 | ||
| CN531555.1 | - | 5 | 689 | |||
| DV660388.1 | - | 1 | 151 | |||

Rnf20
Genomic Map