siRNA Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183015.3 | NP_898836.2 | 4 | 282 | ||
| GenBank mRNA | AJ555464.1 | - | 4 | 283 | ||
| AJ555465.1 | - | 4 | 267 | |||
| BC132411.1 | - | 3 | 236 | 40130688 | ||
| BC138355.1 | - | 3 | 236 | 8861375 | ||

Ccnb3
Genomic Map