siRNA Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183015.3 | NP_898836.2 | 7 | 805 | ||
| GenBank mRNA | AJ555464.1 | - | 7 | 806 | ||
| BC132411.1 | - | 6 | 759 | 40130688 | ||
| BC138355.1 | - | 6 | 759 | 8861375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnb3| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183015.3 | NP_898836.2 | 7 | 805 | ||
| GenBank mRNA | AJ555464.1 | - | 7 | 806 | ||
| BC132411.1 | - | 6 | 759 | 40130688 | ||
| BC138355.1 | - | 6 | 759 | 8861375 | ||