siRNA Details
| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290383.1 | NP_001277312.1 | 16 | 2597 | ||
| NM_194055.3 | NP_918944.2 | 16 | 2600 | |||
| GenBank mRNA | AK140848.1 | - | 16 | 2598 | ||
| BC031468.1 | - | 12 | 1630 | 4037840 | ||
| BC059280.1 | - | 12 | 1640 | |||
| CF161102.1 | - | 1 | 58 | |||

Esrp1
Genomic Map