siRNA Details
Gene Symbol: NTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006181.2 | NP_006172.1 | 1 | 550 | ||
GenBank mRNA | U86759.1 | - | 1 | 550 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
