siRNA Details
| Gene Symbol: Fam120c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198105.2 | NP_932773.2 | 2 | 935 | ||
| GenBank mRNA | AK030477.1 | - | 2 | 984 | ||
| AK136727.1 | - | 2 | 931 | |||
| AK156920.1 | - | 2 | 936 | |||
| AK166381.1 | - | 2 | 915 | |||
| AY150023.1 | - | 2 | 939 | |||
| AY150024.1 | - | 2 | 939 | |||
| BC137973.1 | - | 2 | 935 | 8860992 | ||
| BC171940.1 | - | 2 | 935 | 9054226 | ||

Fam120c
Genomic Map