siRNA Details
| Gene Symbol: Rhox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198598.2 | NP_941000.1 | 1 | 450 | ||
| GenBank mRNA | BC049711.1 | - | 1 | 452 | 6741333 | |
| DQ058648.1 | - | 1 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox11| Gene Symbol: Rhox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198598.2 | NP_941000.1 | 1 | 450 | ||
| GenBank mRNA | BC049711.1 | - | 1 | 452 | 6741333 | |
| DQ058648.1 | - | 1 | 450 | |||