siRNA Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199011.1 | NP_950176.1 | 1 | 208 | ||
| GenBank mRNA | AK048764.1 | - | 1 | 250 | ||
| AK132151.1 | - | 1 | 251 | |||
| AK170840.1 | - | 1 | 229 | |||
| BC062929.1 | - | 1 | 208 | 6855058 | ||
| BC066012.1 | - | 1 | 231 | |||

Dgkq
Genomic Map