siRNA Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199011.1 | NP_950176.1 | 8 | 999 | ||
| GenBank mRNA | AK043933.1 | - | 849 | |||
| AK048764.1 | - | 8 | 1041 | |||
| AK132151.1 | - | 8 | 996 | |||
| AK170840.1 | - | 7 | 1095 | |||
| BC062929.1 | - | 8 | 999 | 6855058 | ||
| BC066012.1 | - | 8 | 976 | |||

Dgkq
Genomic Map