siRNA Details
| Gene Symbol: Gpr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199058.1 | NP_951013.1 | 1 | 929 | ||
| GenBank mRNA | AK139367.1 | - | 2 | 1108 | ||
| AY064488.1 | - | 1 | 929 | |||
| BC111861.1 | - | 1 | 918 | |||
More Information
Set Membership: |
|

Gpr6
Genomic Map