siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199198.2 | NP_954668.2 | 2, 3 | 454 | ||
| GenBank mRNA | AK040235.1 | - | 2, 3 | 406 | ||
| AK045028.1 | - | 2, 3 | 435 | |||
| AK154045.1 | - | 2, 3 | 418 | |||
| BC013700.1 | - | 2, 3 | 407 | |||
| BC064018.1 | - | 2, 3 | 381 | 3980699 | ||
| BU706947.1 | - | 2, 3 | 195 | |||

Hdac10
Genomic Map