siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199198.2 | NP_954668.2 | 15 | 1719 | ||
| GenBank mRNA | AK040235.1 | - | 15 | 1682 | ||
| AK045028.1 | - | 14 | 1930 | |||
| AK154045.1 | - | 14 | 1614 | |||
| BC013700.1 | - | 15 | 1683 | |||
| BC064018.1 | - | 15 | 1646 | 3980699 | ||

Hdac10
Genomic Map