siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199198.2 | NP_954668.2 | 19 | 2189 | ||
| GenBank mRNA | AK040235.1 | - | 18 | 2430 | ||
| AK045028.1 | - | 17 | 2678 | |||
| AK154045.1 | - | 17 | 2362 | |||
| BC013700.1 | - | 19 | 2153 | |||
| BC064018.1 | - | 19 | 2116 | 3980699 | ||

Hdac10
Genomic Map