siRNA Details
| Gene Symbol: Tmem199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199199.3 | NP_954669.2 | 6 | 1167 | ||
| GenBank mRNA | AK160676.1 | - | 1 | 573 | ||
| BC050977.1 | - | 6 | 1130 | |||
| BC064015.1 | - | 2 | 943 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem199| Gene Symbol: Tmem199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199199.3 | NP_954669.2 | 6 | 1167 | ||
| GenBank mRNA | AK160676.1 | - | 1 | 573 | ||
| BC050977.1 | - | 6 | 1130 | |||
| BC064015.1 | - | 2 | 943 | |||