siRNA Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB028272.1 | - | 2 | 734 | ||
| AK002290.1 | - | 2 | 628 | |||
| AK135947.1 | - | 1 | 138 | |||
| AK146657.1 | - | 2 | 696 | |||
| AK158530.1 | - | 2 | 593 | |||
| AK160924.1 | - | 2 | 630 | |||
| AK164392.1 | - | 2 | 659 | |||
| AK167718.1 | - | 2 | 633 | |||
| AK168220.1 | - | 2 | 647 | |||
| AK168291.1 | - | 2 | 647 | |||
| BC012962.1 | - | 2 | 632 | 4188579 | ||

Dnajb1
Genomic Map