siRNA Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018855.2 | NP_061343.2 | 8 | 1083 | ||
| GenBank mRNA | AK140135.1 | - | 8 | 1062 | ||
| AY692443.1 | - | 8 | 1082 | |||
| BC003779.1 | - | 8 | 998 | 3592695 | ||
| U19859.1 | - | 8 | 1104 | |||

Gas8
Genomic Map