siRNA Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018855.2 | NP_061343.2 | 4 | 591 | ||
| GenBank mRNA | AK140135.1 | - | 4 | 570 | ||
| AY692443.1 | - | 4 | 590 | |||
| BC003779.1 | - | 4 | 506 | 3592695 | ||
| DQ189098.1 | - | 3 | 487 | |||
| U19859.1 | - | 4 | 612 | |||

Gas8
Genomic Map