siRNA Details
| Gene Symbol: Herpud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020586.2 | NP_065611.1 | 1 | 939 | ||
| GenBank mRNA | AB041580.1 | - | 1 | 734 | ||
| AK019862.1 | - | 1 | 884 | |||
| AK043046.1 | - | 1 | 803 | |||
| AK135935.1 | - | 1 | 940 | |||
| AK164769.1 | - | 1 | 856 | |||
| BC029691.1 | - | 1 | 157 | 5375603 | ||
| BC043693.1 | - | 1 | 978 | 5150485 | ||

Herpud2
Genomic Map