siRNA Details
| Gene Symbol: Dnaja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135112.1 | NP_001128584.1 | 5 | 804 | ||
| NM_023646.4 | NP_076135.3 | 5 | 804 | |||
| GenBank mRNA | AF325535.1 | - | 5 | 845 | ||
| AK004575.1 | - | 5 | 754 | |||
| AK004910.1 | - | 5 | 756 | |||
| AK031250.1 | - | 5 | 761 | |||
| AK167511.1 | - | 5 | 766 | |||
| AY009320.1 | - | 5 | 845 | |||
| BC003920.1 | - | 5 | 737 | |||
| BC027240.1 | - | 5 | 766 | 2654866 | ||

Dnaja3
Genomic Map