siRNA Details
Gene Symbol: Tnfrsf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023680.3 | NP_076169.2 | 7 | 1744 | ||
GenBank mRNA | AK012838.1 | - | 7 | 1745 | ||
BC076592.1 | - | 6 | 1711 |
Gene Symbol: Tnfrsf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170037.1 | - | 6 | 1722 |
