siRNA Details
| Gene Symbol: Paqr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023824.3 | NP_076313.1 | 1 | 435 | ||
| GenBank mRNA | AB041564.1 | - | 1 | 356 | ||
| AK005142.1 | - | 1 | 458 | |||
| AK033625.1 | - | 1 | 453 | |||
| AK044092.1 | - | 1 | 255 | |||
| AK044486.1 | - | 1 | 458 | |||
| AK152676.1 | - | 1 | 434 | |||
| AY424293.1 | - | 1 | 386 | |||
| BC011185.1 | - | 1 | 435 | 4196033 | ||
| BC026921.1 | - | 1 | 386 | 4500871 | ||

Paqr4
Genomic Map