siRNA Details
| Gene Symbol: 9530002B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023865.3 | NP_076354.1 | 1 | 206 | ||
| GenBank mRNA | AF319955.1 | - | 1 | 191 | ||
| AK020534.1 | - | 1 | 156 | |||
| AY077734.1 | - | 1 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530002B09Rik| Gene Symbol: 9530002B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023865.3 | NP_076354.1 | 1 | 206 | ||
| GenBank mRNA | AF319955.1 | - | 1 | 191 | ||
| AK020534.1 | - | 1 | 156 | |||
| AY077734.1 | - | 1 | 185 | |||