siRNA Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023912.2 | NP_076401.1 | 5 | 885 | ||
| GenBank mRNA | AF276514.1 | - | 5 | 885 | ||
| AK019240.1 | - | 5 | 878 | |||
| AK134356.1 | - | 5 | 884 | |||
| AK144134.1 | - | 5 | 871 | |||
| AK145869.1 | - | 5 | 905 | |||
| AK161813.1 | - | 5 | 884 | |||
| BC034519.1 | - | 5 | 871 | 3672524 | ||
| EF437375.1 | - | 1 | 64 | |||
| HM856325.1 | - | 5 | 885 | |||
| HM856326.1 | - | 5 | 885 | |||

Scyl1
Genomic Map