siRNA Details
| Gene Symbol: Spaca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024279.2 | NP_077241.1 | 6 | 518 | ||
| GenBank mRNA | AK018926.1 | - | 6 | 519 | ||
| BC048522.1 | - | 6 | 514 | 6741699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spaca7| Gene Symbol: Spaca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024279.2 | NP_077241.1 | 6 | 518 | ||
| GenBank mRNA | AK018926.1 | - | 6 | 519 | ||
| BC048522.1 | - | 6 | 514 | 6741699 | ||