siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 5 | 1579 | ||
| GenBank mRNA | AK020014.1 | - | 5 | 1464 | ||
| AK029574.1 | - | 6 | 1654 | |||
| AK049579.1 | - | 5 | 1348 | |||
| AK146492.1 | - | 5 | 1312 | |||
| AK160733.1 | - | 5 | 1168 | |||
| BC046816.1 | - | 5 | 1421 | 5702476 | ||

Desi2
Genomic Map