siRNA Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024475.5 | NP_077795.2 | 10 | 1078 | ||
| GenBank mRNA | AK043952.1 | - | 10 | 985 | ||
| AK050031.1 | - | 10 | 989 | |||
| AK147744.1 | - | 10 | 1026 | |||
| AK153998.1 | - | 10 | 990 | |||
| BC002236.1 | - | 3 | 352 | |||
| BC056652.1 | - | 6 | 579 | |||
| BC083331.1 | - | 8 | 739 | |||
| BC085111.1 | - | 9 | 981 | 30735741 | ||
| BC094051.1 | - | 4 | 384 | |||
| BC106093.1 | - | 9 | 955 | 5098953 | ||

Ublcp1
Genomic Map