siRNA Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028221.4 | NP_082497.2 | 8 | 1225 | ||
| GenBank mRNA | AK033988.1 | - | 8 | 1156 | ||
| AK050704.1 | - | 8 | 1160 | |||
| AK151603.1 | - | 8 | 1179 | |||
| BC016088.1 | - | 7 | 965 | 4503452 | ||
| BC023158.1 | - | 8 | 1064 | 4946201 | ||
| BC025472.1 | - | 8 | 1201 | 5353148 | ||
| BC027060.1 | - | 8 | 1187 | 5358022 | ||
| BC027818.1 | - | 7 | 993 | 5320967 | ||
| BC040818.1 | - | 1178 | 4194531 | |||

Fam192a
Genomic Map