siRNA Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028874.2 | NP_083150.1 | 1 | 547 | ||
| GenBank mRNA | AK122231.1 | - | 1 | 768 | ||
| AK163810.1 | - | 1 | 776 | |||
| BC051164.1 | - | 1 | 768 | |||
| BC063262.1 | - | 1 | 547 | 6405897 | ||

Snx19
Genomic Map