siRNA Details
| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028889.2 | NP_083165.1 | 4 | 1330 | ||
| GenBank mRNA | AK016491.1 | - | 4 | 1330 | ||
| AK045911.1 | - | 5 | 1207 | |||
| AK144175.1 | - | 4 | 1333 | |||
| AK171846.1 | - | 4 | 1332 | |||
| BC019531.1 | - | 4 | 1316 | 4459208 | ||
| BC085088.1 | - | 4 | 1315 | 6467625 | ||

Efhd1
Genomic Map