siRNA Details
| Gene Symbol: Rab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029576.3 | NP_083852.1 | 6 | 929 | ||
| GenBank mRNA | AK003609.1 | - | 6 | 930 | ||
| AK042020.1 | - | 6 | 931 | |||
| AK156726.1 | - | 6 | 952 | |||
| BC016408.1 | - | 6 | 924 | 4242206 | ||

Rab1b
Genomic Map