siRNA Details
| Gene Symbol: 1700018B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029597.1 | NP_083873.1 | 7 | 956 | ||
| GenBank mRNA | AK006079.1 | - | 7 | 956 | ||
| AK006999.1 | - | 7 | 932 | |||
| BC147528.1 | - | 7 | 871 | 9056520 | ||
| BC147542.1 | - | 7 | 871 | 9056533 | ||
| BC172077.1 | - | 7 | 879 | |||
| BC172082.1 | - | 7 | 875 | |||

1700018B08Rik
Genomic Map