siRNA Details
| Gene Symbol: 1700018B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029597.1 | NP_083873.1 | 7 | 1044 | ||
| GenBank mRNA | AK006079.1 | - | 7 | 1044 | ||
| AK006999.1 | - | 7 | 1020 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700018B08Rik| Gene Symbol: 1700018B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029597.1 | NP_083873.1 | 7 | 1044 | ||
| GenBank mRNA | AK006079.1 | - | 7 | 1044 | ||
| AK006999.1 | - | 7 | 1020 | |||